Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, Fusion Flux Additive (Crystalline/For Sample Prepn.)
SDP

Catalog No. ff0910 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FF-0910 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FF-0910 Supplier SPEX CertiPrep Supplier No. FF0910

Specifications

Packaging Poly Bottle
Physical Form Crystalline
Quantity 500 g
Chemical Name or Material Sodium Nitrate, Fusion Flux Additive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.