Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, Granular, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606667 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-667 2.5 kg Poly Pail
18-606-668 12 kg Poly Pail
18-606-669 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-667 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO1832.5KG
Add to Cart
Add to Cart

SO183, 7631-99-4, NaNO3

Spectrum™ Chemical Sodium Nitrate, Granular, FCC is used as curing agent and food preservative that creates a distinct flavor, controls lipid oxidation, and acts as an antimicrobial. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 99 to 100.5%
Grade FCC
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.