Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate (Granular/Laboratory), Fisher Chemical

Catalog No. S3423 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3 kg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S342-3 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S342-3

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Boiling Point 380°C
Melting Point 306°C
Color White
pH 5.5 to 8
Physical Form Crystalline Solid
Quantity 3 kg
Formula Weight 84.99
Grade Laboratory
Identification Pass Test
Packaging Glass Bottle
Chemical Name or Material Sodium Nitrate
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. Irritating to eyes, respiratory system and skin. May cause cancer. Wear personal protective equipment. Keep away from clothing and other combustible materials. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:2
Flammability:1
Instability:3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.