Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Nitrate, puriss. p.a. ACS Reagent, Solstice

Catalog No. 60045955 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-045-955 6 x 500 g
60-014-99 100 g
60-015-01 500 g
60-015-00 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-045-955 Supplier Solstice Supplier No. 314406X500G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO,≥99.5%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-99-4
Molecular Formula NNaO3
Molecular Weight (g/mol) 84.99
MDL Number MFCD00011119
InChI Key VWDWKYIASSYTQR-UHFFFAOYSA-N
Synonym sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
PubChem CID 24268
ChEBI CHEBI:63005
IUPAC Name sodium;nitrate
SMILES [Na+].[O-][N+]([O-])=O

Specifications

Quantity 6 x 500 g
Linear Formula NaNO3
UN Number UN1498
Formula Weight 84.99
Percent Purity ≥99.5%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Nitrate, Reag. ISO
Hazard Statement P220-P305 + P351 + P338
Precautionary Statement H272-H319
DOTInformation 5.1 / PGIII
EINECSNumber 231-554-3
RTECSNumber WC5600000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.