Learn More
GFS Chemicals Inc SODIUM NITRITE, SOLUTION, 0.1 M | 500 ML | GFS CHEMICALS, INC. Item #: 25381 | Bulk #: 2139
SDP
Catalog No. NC9004455
Encompass_Preferred
SODIUM NITRITE, SOLUTION, 0.1 M | Chemical Formula : NaNO!2 | CAS # : 7632-00-0 | Specifications : Concentration (M) 0.0998 - 0.1002 M
Catalog No. NC9004455 Supplier GFS Chemicals Inc Supplier No. 25381
SODIUM NITRITE, SOLUTION, 0.1 M | Chemical Formula : NaNO!2 | CAS # : 7632-00-0 | Specifications : Concentration (M) 0.0998 - 0.1002 M