Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium nitrite, Analytical Standard, 98.0-102.0% (RT), MilliporeSigma™ Supelco™

Catalog No. 111019681 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9681 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-101-9681 Supplier MilliporeSigma Supelco Supplier No. 673981G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sodium nitrite exhibits antioxidant property and is commonly used as a preservative to prevent the chemical and microbiological deterioration of food products.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7632-00-0
Physical Form Neat
Linear Formula NaNO2
MDL Number MFCD00011118
Melting Point 271°C
Synonym E250
Quantity 1 g
Formula Weight 69.00
Percent Purity 98.0 to 102.0% (RT)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.