Learn More
ALADDIN SCIENTIFIC CORPORATION Sodium nitroferricyanide dihyd, 25g
Catalog No. 5000082556
Encompass_Preferred
Sodium nitroprusside (SNP) Dihydrate is a nitric oxide donor/releaser and potent vasodilator which has been reported to reduce lipid peroxidation-induced membrane damage and is believed to exert its vasodilating activity through the vascular activation of nitric oxide SNP is useful for quantitative ammonium analysis as well as chromatographic detection of cystine-containing peptides It has also been reported to be beneficial in maintaining post-thaw sperm viability and motion for up to 6 hours which has been attributed to the reduction of lipid peroxidative damage to the sperm membrane substances
Catalog No. 50-000-82556 Supplier ALADDIN SCIENTIFIC CORPORATION Supplier No. S11075325G
Sodium nitroprusside (SNP) Dihydrate is a nitric oxide donor/releaser and potent vasodilator which has been reported to reduce lipid peroxidation-induced membrane damage and is believed to exert its vasodilating activity through the vascular activation of nitric oxide SNP is useful for quantitative ammonium analysis as well as chromatographic detection of cystine-containing peptides It has also been reported to be beneficial in maintaining post-thaw sperm viability and motion for up to 6 hours which has been attributed to the reduction of lipid peroxidative damage to the sperm membrane substances