Learn More
Medchemexpress LLC HY-B0564 100mg Medchemexpress, Sodium nitroprusside CAS:14402-89-2 Purity:>98%
SDP
Catalog No. 502082600
Encompass_Preferred
Medchemexpress, HY-B0564 100mg Sodium nitroprusside CAS:14402-89-2 Sodium nitroprusside (Ro 21-2498) is a potent vasodilator working through releasing NO spontaneously in blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2600 Supplier Medchemexpress LLC Supplier No. HYB0564100MG
Medchemexpress, HY-B0564 100mg Sodium nitroprusside CAS:14402-89-2 Sodium nitroprusside (Ro 21-2498) is a potent vasodilator working through releasing NO spontaneously in blood. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.