Learn More
Medchemexpress LLC Sodium nonanoate 10g
SDP
Catalog No. 5000205075
Encompass_Preferred
Sodium nonanoate is a salt-like compound commonly used as a surfactant or emulsifier in a variety of industrial applications including the production of detergents soaps and personal care products In addition it can be used as an intermediate in the synthesis of various organic compounds including pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes especially in the manufacture of textiles and paper products
Catalog No. 50-002-05075 Supplier Medchemexpress LLC Supplier No. HYW12737510G
Sodium nonanoate is a salt-like compound commonly used as a surfactant or emulsifier in a variety of industrial applications including the production of detergents soaps and personal care products In addition it can be used as an intermediate in the synthesis of various organic compounds including pharmaceuticals and agrochemicals Its unique chemical properties make it an important ingredient in a variety of industrial processes especially in the manufacture of textiles and paper products