Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium octadecyl sulfate | 1120-04-3 | MFCD00007469 | 98.0% | 372.54 g/mol | C18H37NaO4S | 1 G
SDP

Catalog No. 5000281860
Encompass_Preferred

Sodium octadecyl sulfate is an anionic surfactant and biochemical reagent supplied as a white to off-white solid for research use. It is commonly used as a surfactant and formulation component in biochemical assays, sample preparation, and in vivo formulations, and includes solubility and storage guidance for laboratory handling.

  • High purity (98.0%).
  • White to off-white solid physical form.
  • Soluble in DMSO; warming and sonication improve dissolution.
  • Formulatable for in vivo use in 10% DMSO/90% corn oil.
  • Recommended storage: sealed at 4°C; stock solutions -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Catalog No. 50-002-81860 Supplier Medchemexpress LLC Supplier No. HYW2761641G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium octadecyl sulfate is an anionic surfactant and biochemical reagent supplied as a white to off-white solid for research use. It is commonly used as a surfactant and formulation component in biochemical assays, sample preparation, and in vivo formulations, and includes solubility and storage guidance for laboratory handling.

  • High purity (98.0%).
  • White to off-white solid physical form.
  • Soluble in DMSO; warming and sonication improve dissolution.
  • Formulatable for in vivo use in 10% DMSO/90% corn oil.
  • Recommended storage: sealed at 4°C; stock solutions -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.