Learn More
Medchemexpress LLC Sodium octadecyl sulfate | 1120-04-3 | MFCD00007469 | 98.0% | 372.54 g/mol | C18H37NaO4S | 250 MG
SDP
Supplier:  Medchemexpress LLC HYW276164250MG
Encompass_Preferred
Sodium octadecyl sulfate is an anionic surfactant and biochemical reagent used in life-science research. It serves as the sodium salt of octadecyl sulfate and is employed in applications that require anionic detergents, such as sample preparation, membrane solubilization, and biochemical assays. The substance is supplied for laboratory research use and is characterized by a reported purity of 98.0%.
  • High purity (98.0%) suitable for analytical and research applications.
  • Anionic surfactant for protein denaturation and membrane solubilization.
  • Useful in formulation of biochemical assays and sample preparation.
  • Available in small lab pack sizes for bench-scale experiments.
  • Provided with datasheet, COA, and SDS for handling and quality assurance.
Catalog No. 50-002-82313