Learn More
Medchemexpress LLC Sodium octadecyl sulfate (sodium stearyl sulfate) | 1120-04-3 | MFCD00007469 | 98.0% | 372.54 g/mol | C18H37NaO4S | 500 MG
SDP
Catalog No. 5000282436
Encompass_Preferred
Sodium octadecyl sulfate (sodium stearyl sulfate) is an anionic surfactant and biochemical reagent used in life-science research for formulation work, protein solubilization, and analytical assays. It is provided with purity specifications and safety documentation for laboratory handling and small-scale experimental use.
  • Anionic surfactant for solubilization and emulsification.
  • High purity (98.0%) suitable for research applications.
  • Supplied in small laboratory pack sizes for convenience.
  • SDS available for safe handling and storage guidance.
Catalog No. 50-002-82436 Supplier Medchemexpress LLC Supplier No. HYW276164500MG
Sodium octadecyl sulfate (sodium stearyl sulfate) is an anionic surfactant and biochemical reagent used in life-science research for formulation work, protein solubilization, and analytical assays. It is provided with purity specifications and safety documentation for laboratory handling and small-scale experimental use.
  • Anionic surfactant for solubilization and emulsification.
  • High purity (98.0%) suitable for research applications.
  • Supplied in small laboratory pack sizes for convenience.
  • SDS available for safe handling and storage guidance.