Learn More
Medchemexpress LLC Sodium octane-1-sulfonate | 5324-84-5 | 99.9% | 216.27 | 50 G
SDP
Catalog No. 5000452179
Encompass_Preferred
Sodium octane-1-sulfonate is an ion-pairing agent that can be used as a buffer in biochemical research and HPLC. It is for research use only.
  • Used as a buffer in biochemical research
  • Suitable for HPLC applications
  • For research use only
Catalog No. 50-004-52179 Supplier Medchemexpress LLC Supplier No. HYW00860050G
Sodium octane-1-sulfonate is an ion-pairing agent that can be used as a buffer in biochemical research and HPLC. It is for research use only.
  • Used as a buffer in biochemical research
  • Suitable for HPLC applications
  • For research use only