Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium octanoate, 1984-06-1, MFCD00058511, 100 g
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C5038100G
Encompass
Linear Formula: CH3(CH2)6COONa, Molecular Weight: 166.19, ≥99% (capillary GC), Synonym: Caprylic acid sodium salt, Octanoic acid sodium salt, Sodium caprylate.
Catalog No. NC0705955