Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium octyl sulfate 9525G
Catalog No. 502907754
Encompass
Sodium octyl sulfate is an anionic detergent and ion pairing reagent that is used in HPLC
Catalog No. 50-290-7754 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. O400325G
Sodium octyl sulfate is an anionic detergent and ion pairing reagent that is used in HPLC