Learn More
Medchemexpress LLC Sodium oleate | 143-19-1 | 304.44 | 25 G
SDP
Catalog No. 5000389575
Encompass_Preferred
Sodium oleate is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid (FA) in human adipocytes and other tissues. This compound promotes cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation. Conversely, it inhibits cancer cell growth and survival in low metastatic carcinoma cells, such as gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines.
  • Abundant monounsaturated fatty acid sodium
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration in high metastatic cancer cells
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells
  • For research use only
Catalog No. 50-003-89575 Supplier Medchemexpress LLC Supplier No. 25GHYN1446B
Sodium oleate is an abundant monounsaturated fatty acid sodium and a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid (FA) in human adipocytes and other tissues. This compound promotes cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation. Conversely, it inhibits cancer cell growth and survival in low metastatic carcinoma cells, such as gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines.
  • Abundant monounsaturated fatty acid sodium
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration in high metastatic cancer cells
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells
  • For research use only