Learn More
Medchemexpress LLC Sodium oleate | 143-19-1 | 304.44 | 5 G
SDP
Catalog No. 5000414675
Encompass_Preferred
Sodium oleate is an abundant monounsaturated fatty acid sodium that acts as a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid in human adipocytes and other tissues. It can promote cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation, and it inhibits cancer cell growth in low metastatic carcinoma cells.
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration
  • Inhibits cancer cell growth
Catalog No. 50-004-14675 Supplier Medchemexpress LLC Supplier No. HYN1446B5G
Sodium oleate is an abundant monounsaturated fatty acid sodium that acts as a Na+/K+ ATPase activator. It is the most common monounsaturated fatty acid in human adipocytes and other tissues. It can promote cell proliferation and migration in high metastatic cancer cells by enhancing β-oxidation, and it inhibits cancer cell growth in low metastatic carcinoma cells.
  • Na+/K+ ATPase activator
  • Promotes cell proliferation and migration
  • Inhibits cancer cell growth