Learn More
Medchemexpress LLC Sodium oleate | 143-19-1 | 304.44 | 500 MG
SDP
Catalog No. 5000415705
Encompass_Preferred
Sodium oleate, also known as Oleic acid sodium, is a plentiful monounsaturated fatty acid sodium that functions as a Na+/K+ ATPase activator. Oleic acid is the most prevalent monounsaturated fatty acid found in human adipocytes and other tissues.
  • Promotes cell proliferation and migration in highly metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation.
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells, such as gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines.
  • Acts as a Na+/K+ ATPase activator.
Catalog No. 50-004-15705 Supplier Medchemexpress LLC Supplier No. HYN1446B500MG
Sodium oleate, also known as Oleic acid sodium, is a plentiful monounsaturated fatty acid sodium that functions as a Na+/K+ ATPase activator. Oleic acid is the most prevalent monounsaturated fatty acid found in human adipocytes and other tissues.
  • Promotes cell proliferation and migration in highly metastatic cancer cells by enhancing β-oxidation mediated by AMPK activation.
  • Inhibits cancer cell growth and survival in low metastatic carcinoma cells, such as gastric carcinoma SGC7901 and breast carcinoma MCF-7 cell lines.
  • Acts as a Na+/K+ ATPase activator.