Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oleate, Spectrum™ Chemical
SDP

Catalog No. 18621458 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-458 50 kg
18-621-454 25 g
18-621-455 100 g
18-621-456 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-458 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S160550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 143-19-1
Molecular Weight (g/mol) 304.44 g/mol

Specifications

Packaging Drum
Quantity 50 kg
Physical Form Powder or Waxy Solid
Chemical Name or Material Sodium Oleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.