Learn More
New England Biolabs, Inc. Sodium Orthovanadate (Vanadate) – 1 ml
SDP
Catalog No. 50994911
Encompass
Sodium Orthovanadate (Vanadate) is a general competitive inhibitor for protein phosphotyrosyl phosphatases. The inhibition by Sodium Orthovanadate is reversible upon the addition of EDTA or by dilution.
Catalog No. 50-994-911 Supplier New England Biolabs, Inc. Supplier No. P0758S
Sodium Orthovanadate (Vanadate) is a general competitive inhibitor for protein phosphotyrosyl phosphatases. The inhibition by Sodium Orthovanadate is reversible upon the addition of EDTA or by dilution.