Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oxalate, 0.0250 M (M/40), 0.0500 N (N/20), Ricca Chemical
SDP

Catalog No. 7499532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7499-532 1 L Natural Poly Bottle
7499-516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 7499-532 Supplier Ricca Chemical Company Supplier No. 7499.532
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5 , 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color Colorless
CAS Min % 98.0, 0.33
CAS Max % 100.0, 0.34
Physical Form Liquid
Quantity 1 L
Grade Laboratory
Concentration or Composition (by Analyte or Components) 0.0250 Molar (M/40), 0.0500 Normal (N/20)
Packaging Natural Poly Bottle
Chemical Name or Material Sodium Oxalate
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.