Learn More
Alkali Scientific Sodium Oxalate, 100 Grams
SDP
Catalog No. 5000659741
Encompass_Preferred
Sodium Oxalate is a high-purity oxalate salt used in laboratory research applications, primarily in the preparation of various chemical reagents. As a reagent, it plays an important role in oxalate precipitation reactions and serves as a key intermediate in organic synthesis. It is also commonly used in quantitative analysis to determine calcium content in solutions. This 100-gram package is ideal for use in analytical chemistry, particularly for studies involving calcium oxalate and other inorganic compounds. Sodium oxalate is also utilized in the synthesis of oxalate complexes and as a buffering agent in some chemical processes.
Catalog No. 50-006-59741 Supplier Alkali Scientific Supplier No. IUM001CP
Sodium Oxalate is a high-purity oxalate salt used in laboratory research applications, primarily in the preparation of various chemical reagents. As a reagent, it plays an important role in oxalate precipitation reactions and serves as a key intermediate in organic synthesis. It is also commonly used in quantitative analysis to determine calcium content in solutions. This 100-gram package is ideal for use in analytical chemistry, particularly for studies involving calcium oxalate and other inorganic compounds. Sodium oxalate is also utilized in the synthesis of oxalate complexes and as a buffering agent in some chemical processes.