Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oxalate, ACS Reagent, Reagents

Catalog No. 01337706
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-706 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-337-706 Supplier Reagents Holdings Llc Supplier No. C1322000100C2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Oxalate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym Disodium Ethanedioic Acid
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Percent Purity ≥99.0%
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.34
Chemical Name or Material Sodium Oxalate, ACS Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.