Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium oxalate ACS reagent500G
Catalog No. 502899296
Encompass
Sodium oxalate is a water soluble inorganic salt mainly used for the standardization of permanganate SOLUTIONon during titration.
Catalog No. 50-289-9296 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 223433500G
Sodium oxalate is a water soluble inorganic salt mainly used for the standardization of permanganate SOLUTIONon during titration.