Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oxalate (Certified ACS), Fisher Chemical

Catalog No. S487500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S487-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S487-500
This item is not returnable. View return policy

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Melting Point 250°C
Color White
Density 2.34g/mL
pH 8
Loss on Drying 0.01% max.
Assay Percent Range ≥99.5 %
Packaging Glass Bottle
Linear Formula NaO2CCO2Na
Quantity 500 g
UN Number 2811
Solubility Information 0.005% max. Insoluble Matter
Formula Weight 134
Percent Purity ≥99.5%
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material Sodium Oxalate
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 6.1: Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.