Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oxalate Solution, 0.05M, Solstice

Catalog No. 6002616 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-026-16 1 L
60-026-17 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-026-16 Supplier Solstice Supplier No. 352401L
Add to Cart
Add to Cart

Volumetric, 0.05 M (COO)2Na2

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-76-0
Molecular Formula C2Na2O4
Molecular Weight (g/mol) 134.00
MDL Number MFCD00012465
InChI Key ZNCPFRVNHGOPAG-UHFFFAOYSA-L
Synonym sodium oxalate, disodium oxalate, natriumoxalat, ethanedioic acid, disodium salt, oxalic acid, disodium salt, natriumoxalat german, stavelan sodny czech, oxalic acid disodium salt, unii-7u0v68lt9x, ethanedioic acid disodium salt
PubChem CID 6125
IUPAC Name disodium;oxalate
SMILES [Na+].[Na+].[O-]C(=O)C([O-])=O

Specifications

Quantity 1 L
Linear Formula NaOOCCOONa
UN Number NONH for all modes of transport
Beilstein 3631622
Formula Weight 134g/mol
Chemical Name or Material Sodium oxalate solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.