Learn More
Cayman Chemical Sodium Oxamate, 565-73-1, 5 g
SDP
Supplier:  Cayman Chemical 190575
Encompass_Preferred
Molecular Formula C2H2NO3 • Na, Purity ≥95%, Formula Weight 111.0, Synonyms: Oxalic Acid monoamide, Oxamic Acid; A derivative of pyruvate that inhibits the conversion of pyruvate to lactate via lactate dehydrogenase, thus disrupting glycolysis; studied as an inhibitor of glycolysis in various tumors.
Catalog No. NC1176295