Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium Oxide

Catalog No. 645613250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
64-561-3250G 250 g
64-561-350G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 64-561-3250G Supplier Sigma Aldrich Supplier No. 645613250G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium oxide is a strong alkaline oxide, commonly used as an active flux in ceramic glazes.Sodium oxide has been used as a coating precursor on organic polymer films during the plasma modification to analyze its antimicrobial properties.

Specifications

CAS 1313-59-3
Quantity 250 g
Molecular Formula Na2O
Linear Formula Na2O
MDL Number MFCD00046201
Synonym Disodium monoxide; Disodium oxide
Molecular Weight (g/mol) 61.98 g/mol
Density 2.27 g/mL at 25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.