Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium p-toluenesulfonate

Catalog No. 152536500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-253-6500G 500 g
15-253-6100G 100 g
15-253-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-253-6500G Supplier Sigma Aldrich Supplier No. 152536500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Sodium p-toluenesulfonate was used as a supporting electrolyte for depositing polypyrrole membranes
  • It was also used as a solute to study the performance of resin particles

Specifications

CAS 657-84-1
Molecular Formula C7H7NaO3S
Linear Formula CH3C6H4SO3Na
MDL Number MFCD00064388
Quantity 500 g
Synonym p-Toluenesulfonic acid sodium salt
Molecular Weight (g/mol) 194.18
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.