Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium (para)periodate, 98%

Catalog No. AC432855000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC432855000 500 g Glass bottle
AC432851000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC432855000 Supplier Thermo Scientific Chemicals Supplier No. 432855000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13940-38-0 ∣ H2INa3O6 ∣ 293.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13940-38-0
Molecular Formula H2INa3O6
Molecular Weight (g/mol) 293.88
MDL Number MFCD00036146
InChI Key DSYMXPNUZLZKOA-UHFFFAOYSA-K
Synonym sodium paraperiodate, sodium para periodate, sodium triparaperiodate, sodium hydrogen periodate, 3na.h2io6, periodic acid trisodium salt, trisodium dihydrogen orthoperiodate, sodium para periodate, commercial, trisodium dihydroxydioxidoiodosylolate, trisodium dihydroxy-trioxido-oxo-$l^ 7-iodane
PubChem CID 9796123
IUPAC Name trisodium;dihydroxy-trioxido-oxo-$l^{7}-iodane
SMILES [Na+].[Na+].[Na+].O[I](O)([O-])([O-])([O-])=O

Specifications

Melting Point >300.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula Na3H2IO6
Solubility Information Solubility in water: slightly soluble
Formula Weight 293.89
Percent Purity 98%
Grade Ungraded
Packaging Glass bottle
Chemical Name or Material Sodium (para)periodate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
In case of major fire and large quantities: Evacuate area.
Fight fire remotely due to the risk of explosion.
IF ON SKIN: Wash with plenty of soap an
EINECSNumber 237-720-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.