Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pentacyanoammineferroate(II) Hydrate 95.0+%, TCI America™
SDP

Catalog No. S005025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S005025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S005025G Supplier TCI America Supplier No. S005025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14099-05-9
MDL Number MFCD00059942
Synonym Pentacyanoammineferroate(II) Sodium Salt, Trisodium Amminepentacyanoferroate(II)

Specifications

Color Yellow
Quantity 25 g
UN Number 1588
Formula Weight 271.94
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Pentacyanoammineferroate(II) Hydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.