Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium pentafluoropropionate

Catalog No. 38172150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-172-150G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 38-172-150G Supplier Sigma Aldrich Supplier No. 38172150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium pentafluoropropionate

Specifications

CAS 378-77-8
Melting Point 226°C to 229°C (dec.) (lit.)
Molecular Formula C3F5NaO2
Linear Formula CF3CF2COONa
MDL Number MFCD00066167
Quantity 100 mg
Synonym Pentafluoropropionic acid sodium salt; Perfluoropropionic acid sodium salt
Molecular Weight (g/mol) 186.01
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.