Learn More
Medchemexpress LLC Sodium pentanesulfonate | 22767-49-3 | MFCD00007541 | ≥98.0% | 174.19 g/mol | C5H11NaO3S | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW016791
Encompass_Preferred
Sodium pentanesulfonate (sodium pentane-1-sulfonate) is an anionic surfactant and biochemical reagent used to enhance solubility of proteins and membrane proteins in biochemical assays. The sodium salt is provided at high purity and is accompanied by safety and quality documentation for laboratory use.
  • Anionic surfactant that enhances protein solubility.
  • High purity suitable for biochemical and analytical assays.
  • Facilitates solubilization of membrane proteins.
  • Supplied with SDS and certificate of analysis.
  • Available in laboratory-scale packaging for experimental use.
Catalog No. 50-003-03093