Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium perborate tetrahydrate 9-11% available Oxygen | 10486-00-7 | MFCD00149231 | 500G
Supplier: Sigma Aldrich Fine Chemicals Biosciences 244120500G
Sodium perborate tetrahydrate 9-11% available Oxygen | Purity: ~6.5-7.5% NaOH basis (titration); ~9-11% active oxygen basis (by Na2S2O3, titration) | Mol Wt: 153.86 | 10486-00-7 | MFCD00149231 | 500G