Learn More
Alkali Scientific Sodium Perchlorate, ACS Grade, 1 Kg
SDP
Catalog No. 5000661782
Encompass_Preferred
Sodium Perchlorate, ACS Grade, is a high-purity inorganic salt used as an oxidizing agent in a variety of chemical reactions. Available in a 1 kg quantity, it is ideal for laboratory applications requiring a stable and reliable source of sodium perchlorate. Commonly used in the synthesis of other chemicals and in various research procedures, sodium perchlorate is widely utilized in the preparation of perchloric acid and in chemical analysis processes. Its oxidizing properties make it valuable for reactions that require high-temperature stability and controlled oxidation conditions. This chemical is suitable for use in both organic and inorganic chemistry applications.
Catalog No. 50-006-61782 Supplier Alkali Scientific Supplier No. UMP003CP
Sodium Perchlorate, ACS Grade, is a high-purity inorganic salt used as an oxidizing agent in a variety of chemical reactions. Available in a 1 kg quantity, it is ideal for laboratory applications requiring a stable and reliable source of sodium perchlorate. Commonly used in the synthesis of other chemicals and in various research procedures, sodium perchlorate is widely utilized in the preparation of perchloric acid and in chemical analysis processes. Its oxidizing properties make it valuable for reactions that require high-temperature stability and controlled oxidation conditions. This chemical is suitable for use in both organic and inorganic chemistry applications.