Learn More
Alkali Scientific Sodium Perchlorate, ACS Grade, 5 Kg
SDP
Catalog No. 5000660611
Encompass_Preferred
Sodium Perchlorate, ACS Grade, is a high-purity oxidizing agent available in a 5 kg quantity, commonly used for laboratory and industrial applications. This inorganic salt is valued for its stability and reliability in a variety of chemical reactions. It is often used in the synthesis of perchloric acid and other related compounds. As an oxidizing agent, sodium perchlorate facilitates oxidation reactions, making it an essential component in laboratory experiments involving high temperatures and controlled chemical environments. It is ideal for research involving oxidizing agents in both organic and inorganic chemistry.
Catalog No. 50-006-60611 Supplier Alkali Scientific Supplier No. UMP004CP
Sodium Perchlorate, ACS Grade, is a high-purity oxidizing agent available in a 5 kg quantity, commonly used for laboratory and industrial applications. This inorganic salt is valued for its stability and reliability in a variety of chemical reactions. It is often used in the synthesis of perchloric acid and other related compounds. As an oxidizing agent, sodium perchlorate facilitates oxidation reactions, making it an essential component in laboratory experiments involving high temperatures and controlled chemical environments. It is ideal for research involving oxidizing agents in both organic and inorganic chemistry.