Learn More
Sodium Perchlorate Buffer 60% and Acetonitrile 40%, LabReady™ Solvent Blend, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Chemical Name or Material | Sodium Perchlorate Buffer 60% and Acetonitrile 40% |
| Name Note | LabReady™ Solvent Blend |
| CAS | 7732-18-5, 75-05-8, 7601-89-0 |
| Boiling Point | 70.56°C |
| CAS Max % | 56.4000%, 29.5000%, 14.1000% |
| Density | 1.0267 g/cm3 |
| Grade | Solvent Blend |
| Packaging | Glass Bottle |
| Quantity | 4 x 4 L |
| Flash Point | 10°C |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.