Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Perchlorate Monohydrate, 98%, ACS Reagent, Solstice

Catalog No. 60045890 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-890 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-045-890 Supplier Solstice Supplier No. 310514500G

missing translation for 'validMainframeMsg'

ACS Reagent, 98%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-3-7791
ChEBI CHEBI:78672

Specifications

Melting Point 469°C
Quantity 500 g
Molecular Formula ClRb
Linear Formula NaClO4 · H2O
MDL Number MFCD00011187
UN Number UN1502
Synonym rubidium chloride, rubidium chloride rbcl, rubidium monochloride, rbcl, rubidium 1+ chloride, rubidium 1+ ion chloride, unii-n3shc5273s, rubidiumchloride, rubidium-chlorid, rubidium-chloride
InChI Key FGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES [Cl-].[Rb+]
IUPAC Name rubidium(1+);chloride
Molecular Weight (g/mol) 120.92
PubChem CID 62683
Formula Weight 140.46g/mol
Percent Purity 98%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.02g/cm3
Chemical Name or Material Sodium Perchlorate Monohydrate
Show More Show Less
Hazard Statement P220
Precautionary Statement H271-H302
DOTInformation 5.1/PGII
EINECSNumber 231-511-9
RTECSNumber SC9850000
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.