Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Perchlorate Monohydrate, purum p.a., ≥98% (AT), Solstice

Catalog No. 6001529 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-29 250 g
60-015-28 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-015-29 Supplier Solstice Supplier No. 71852250G

missing translation for 'validMainframeMsg'

Purum p.a., crystallized,≥98.0% (T)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7-3-7791
Molecular Formula ClH2NaO5
Molecular Weight (g/mol) 140.45
MDL Number MFCD00149164
InChI Key IXGNPUSUVRTQGW-UHFFFAOYSA-M
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
PubChem CID 516899
IUPAC Name sodium;perchlorate;hydrate
SMILES O.[O-]Cl(=O)(=O)=O.[Na+]

Specifications

Quantity 250 g
Linear Formula NaClO4 · H2O
UN Number UN1502
Formula Weight 140.46g/mol
Percent Purity ≥98% (T)
Chemical Name or Material Sodium perchlorate monohydrate
Hazard Statement P220
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.