Learn More
Hach Company Sodium Periodate Powder Pillows for Manganese, 25 mL, pk/100
Catalog No. 501994486 Shop All Hach Company Products
Encompass
For determination of manganese by the periodate oxidation method. EPA approved for reporting wastewater analyses (digestion required). Hach method 8034. Sample size: 25 mL Pack of 100 Powder Pillows.
Catalog No. 50-199-4486 Supplier Hach Company Supplier No. 98499
For determination of manganese by the periodate oxidation method. EPA approved for reporting wastewater analyses (digestion required). Hach method 8034. Sample size: 25 mL Pack of 100 Powder Pillows.