Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium permanganate solution

Catalog No. 519073500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-907-3500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 51-907-3500ML Supplier Sigma Aldrich Supplier No. 519073500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium permanganate solution (NaMnO4) is a water soluble salt solution that can prepared by using manganese oxide with sodium hypochloride. It can be used as an etchant in electronic circuits. It can also be used as a fixing agent in electron microscopy.

Specifications

Boiling Point 100°C
CAS 10101-50-5
Quantity 500 mL
Molecular Formula MnNaO4
Linear Formula NaMnO4
MDL Number MFCD00003481
Synonym Sodium manganate; Sodium permanganate (NaMnO4)
Molecular Weight (g/mol) 141.93
Density 1.391 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.