Learn More
Aqua Solutions Sodium Phenate Reagent APHA & EPA 350.1 for Ammonia (500ML)
SDP
Catalog No. NC9451714
Encompass_Preferred
Sodium Phenate Reagent APHA & EPA 350.1 for Ammonia (500ML)
Catalog No. NC9451714 Supplier Aqua Solutions Supplier No. 8710500ML
Sodium Phenate Reagent APHA & EPA 350.1 for Ammonia (500ML)