Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium phenyl phosphate dibasic dihydrate

Catalog No. P775110G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P775110G 10 g
P7751100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P775110G Supplier Sigma Aldrich Supplier No. P775110G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Sodium phenyl phosphate dibasic dihydrate

Specifications

CAS 66778-08-3
Molecular Formula C6H5Na2O4P -+ 2H2O
Linear Formula C6H5PO4Na2 -+ 2H2O
MDL Number MFCD00149080
Quantity 10 g
Synonym Disodium phenyl phosphate dihydrate; Phenyl phosphate disodium salt
Molecular Weight (g/mol) 254.09
Percent Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.