Learn More
Medchemexpress LLC Sodium phenyl phosphate dihydrate | 66778-08-3 | MFCD00149080 | 100g
SDP
Catalog No. 5000266072
Encompass_Preferred
Sodium phenyl phosphate dihydrate can be used as a buffering agent in biochemical and pharmaceutical formulations Sodium phenyl phosphate dehydrate also acts as a substrate for evaluating phosphataseactivity[1]
Catalog No. 50-002-66072 Supplier Medchemexpress LLC Supplier No. HYZ0290A100G
Sodium phenyl phosphate dihydrate can be used as a buffering agent in biochemical and pharmaceutical formulations Sodium phenyl phosphate dehydrate also acts as a substrate for evaluating phosphataseactivity[1]