Learn More
Medchemexpress LLC Sodium phenyl phosphate (dihydrate) | 66778-08-3 | MFCD00149080 | 50g
SDP
Catalog No. 5000266068
Encompass_Preferred
Sodium phenyl phosphate dihydrate can be used as a buffering agent in biochemical and pharmaceutical formulations Sodium phenyl phosphate dehydrate also acts as a substrate for evaluating phosphataseactivity[1]
Catalog No. 50-002-66068 Supplier Medchemexpress LLC Supplier No. HYZ0290A50G
Sodium phenyl phosphate dihydrate can be used as a buffering agent in biochemical and pharmaceutical formulations Sodium phenyl phosphate dehydrate also acts as a substrate for evaluating phosphataseactivity[1]