Learn More
Active Motif Sodium phenylbutyrate
SDP
Catalog No. NC1838785
Encompass
Active Motif's Phenylbutyrate Na is a histone deacetylase (HDAC) inhibitor that inhibits proliferation, migration and invasion of a number of cancer cell lines. It induces differentiation and apoptosis, and is active in vivo.
Catalog No. NC1838785 Supplier Active Motif Supplier No. 14033
Active Motif's Phenylbutyrate Na is a histone deacetylase (HDAC) inhibitor that inhibits proliferation, migration and invasion of a number of cancer cell lines. It induces differentiation and apoptosis, and is active in vivo.