Learn More
Selleck Chemical LLC Sodium Phenylbutyrate 50mg 1716-12-7 4-PBA, 4-Phenylbutyric acid
SDP
Supplier:  Selleck Chemical LLC S412550MG
Encompass_Preferred
Sodium phenylbutyrate is a salt of 4-phenylbutyrate (4-PBA) or 4-phenylbutyric acid.Sodium phenylbutyrate is a histone deacetylase inhibitor, used to treat urea cycle disorders. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8021