Learn More
Selleck Chemical LLC Sodium phenylpyruvate S3351-100mg
SDP
Catalog No. 503109876
Encompass_Preferred
Sodium phenylpyruvate (Phenylpyruvic acid sodium salt) inhibits amino acid formation and depresses oxygen consumption
Catalog No. 50-310-9876 Supplier Selleck Chemical LLC Supplier No. S3351100MG
Sodium phenylpyruvate (Phenylpyruvic acid sodium salt) inhibits amino acid formation and depresses oxygen consumption