Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium phosphate monobasic meets USP testing specifications, anhydrous | 7558-80-7 | MFCD00003527 | 100G
Catalog No. NC1467065
Encompass
Sodium phosphate monobasic meets USP testing specifications, anhydrous | Purity: 98.0-103.0% (T) | Mol Wt: 119.98 | 7558-80-7 | MFCD00003527 | 100G
Catalog No. NC1467065 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. S2554100G
Sodium phosphate monobasic meets USP testing specifications, anhydrous | Purity: 98.0-103.0% (T) | Mol Wt: 119.98 | 7558-80-7 | MFCD00003527 | 100G