Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Sodium phosphate, 0.5M buffer soln., pH 7.0

Catalog No. AAJ63791AK
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63791AK 250 mL
AAJ63791AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63791AK Supplier Thermo Scientific Chemicals Supplier No. J63791AK
Add to Cart

Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate is widely used in molecular biology, biochemistry, and chromatography. Useful in conjunction with Sodium Phosphate, Monobasic in the preparation of biological buffers. Used in many applications including the purification of antibodies.

Solubility
Fully miscible.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium phosphate buffer soln.
Concentration 0.5 M
pH 7.0
Physical Form Liquid
Quantity 250 mL
MDL Number MFCD00146206
Solubility Information Fully miscible.
The specifications for Sodium phosphate, 0.5M buffer soln., pH 7.0 say to store in cool dry conditions. Does this mean I need to store it in the refrigerator?

Sodium phosphate, 0.5M buffer soln., pH 7.0 can be stored at ambient temperature, 15-25° Celsius. It does not need to be stored in a refrigerator.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.